N,N-Diisopropylethylenediamine 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Aminoethyldiisopropylamine, N,N-Bis(isopropyl)ethane-1,2-diamine
L04765.09
L04765.18
L04765.09EA
1300
CZK
L04765.09
L04765.18
N,N-Diisopropylethylenediamine 97%
N,N-Diisopropylethylenediamine
Purity: 97%
CAS No.: 121-05-1
Molecular Formula: C8H20N2
Formula Weight: 144.26
Flash Point: 54°(129°F)
Refractive Index: 1.4420
Boiling Point: 170°
Beilstein Reference: 1734800
UN No.: 2733
MDL No.: MFCD00015015
Vzorec:
C₈H₂₀N₂ Bod varu: 170 °C Hustota: 0,828 Bod vzplanutí: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015015 CAS číslo: 121-05-1 EINECS: 204-447-4 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...