D-(-)-α-(4-Hydroxyphenyl)glycine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine
L07190.06
L07190.14
L07190.06EA
635
CZK
L07190.06
L07190.14
D-(-)-α-(4-Hydroxyphenyl)glycine ≥98%
D-(-)-α-(4-Hydroxyphenyl)glycine
Purity: 98+%
CAS No.: 22818-40-2
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: ca 240° dec.
Beilstein Reference: 2210998
Vzorec:
C₈H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00004262 CAS číslo: 22818-40-2 EINECS: 245-247-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...