Tisk…

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

673943-25G 673943-5G
SIAL673943-25GEA 4080 CZK
SIAL673943-25G SIAL673943-5G
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Sigma-Aldrich®
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec: C₇H₁₅BO₃
MW: 158,01 g/mol
MDL Number: MFCD03093086
CAS číslo: 1195-66-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR