Tisk…

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Isopropyl pinacolyl borate, 4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane

L17278.06 L17278.14
L17278.06EA 2130 CZK
L17278.06 L17278.14
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98%
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Purity: 98% CAS No.: 61676-62-8 Molecular Formula: C9H19BO3 Formula Weight: 186.06 Flash Point: 42°(107°F) Refractive Index: 1.4090 Boiling Point: 73°/15mm Beilstein Reference: 1906370 UN No.: 1993 MDL No.: MFCD00192241

Vzorec: C₉H₁₉BO₃
MW: 186,06 g/mol
Bod varu: 73 °C (15 mmHg)
Hustota: 0,912
Bod vzplanutí: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192241
CAS číslo: 61676-62-8
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR