Tisk…

3-(2,5-Dimethoxyphenyl)propionic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

H33566.03 H33566.06 H33566.14
H33566.03EA 965 CZK
H33566.03 H33566.06 H33566.14
3-(2,5-Dimethoxyphenyl)propionic acid 96%
3-(2,5-Dimethoxyphenyl)propionic acid

Purity: 96% CAS No.: 10538-49-5 Molecular Formula: C11H14O4 Formula Weight: 210.23 Melting Point: 61-66° MDL No.: MFCD00060325

Vzorec: C₁₁H₁₄O₄
Bod tání: 61…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00060325
CAS číslo: 10538-49-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR