Tisk…

α-Chlorothioanisole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Chloromethyl phenyl sulphide

216631-25G 216631-5G
SIAL216631-25GEA 7220 CZK
SIAL216631-25G SIAL216631-5G
α-Chlorothioanisole, Sigma-Aldrich®
α-Chlorothioanisole
Vzorec: C₇H₇ClS
MDL Number: MFCD00000921
CAS číslo: 7205-91-6
EINECS: 230-579-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR