Tisk…

Rhodium(II) acetate dimer, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

482285-1G 482285-250MG
SIAL482285-1GEA 44790 CZK
SIAL482285-1G SIAL482285-250MG
Rhodium(II) acetate dimer, Sigma-Aldrich®
Rhodium(II) acetate dimer
Vzorec: C₈H₁₂O₈Rh₂
MDL Number: MFCD00003538
CAS číslo: 15956-28-2
EINECS: 240-084-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR