Dinonyl phthalate mixture of isomers 96%
Dodavatel: Thermo Scientific
408770250
408771000
ACRO408770250EA
3100
CZK
ACRO408770250
ACRO408771000
Dinonyl phthalate mixture of isomers 96%
Dinonyl phthalate
Vzorec:
C₂₆H₄₂O₄ Bod varu: 279…287 °C (760 mmHg) Hustota: 0,97 g/cm³ Bod vzplanutí: 216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036237 CAS číslo: 84-76-4 EINECS: 201-560-0 ADR: 9,III |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow-brown Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4820 to 1.4910 (20°C, 589 nm) |
GC | ≥95.0 % (sum of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...