Tisk…

Methyl-3-methoxypropionate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 3-methoxypropionate

M-Clarity™ quality level = MQ200

407429-1L 407429-250ML
SIAL407429-1LEA 4410 CZK
SIAL407429-1L SIAL407429-250ML
Methyl-3-methoxypropionate, Sigma-Aldrich®
Methyl-3-methoxypropionate
Vzorec: C₅H₁₀O₃
MW: 118,1311 g/mol
MDL Number: MFCD00059085
CAS číslo: 3852-09-3
EINECS: 223-358-1
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR