Tisk…

2-Bromopropane-[D₇] (98% D), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Bromopropane-[²H₇]

M-Clarity™ quality level = MQ200

375608-1G 375608-5G
SIAL375608-1GEA 2740 CZK
SIAL375608-1G SIAL375608-5G
2-Bromopropane-[D₇] (98% D), Sigma-Aldrich®
2-Bromopropane-[D₇]
Vzorec: C₃D₇Br
MW: 130,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075610
CAS číslo: 39091-63-9
UN: 2344
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR