Tisk…

N-Fmoc-glycine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

47627-50G-F 47627-5G-F
SIAL47627-50G-FEA 2390 CZK
SIAL47627-50G-F SIAL47627-5G-F
N-Fmoc-glycine, Sigma-Aldrich®
N-Fmoc-glycine
Vzorec: C₁₇H₁₅NO₄
MW: 297.3 g/mol
MDL Number: MFCD00037140
CAS číslo: 29022-11-5
EINECS: 249-373-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR