Tisk…

N-Fmoc-glycine ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

B21050.03 B21050.06
B21050.03EA 642 CZK
B21050.03 B21050.06
N-Fmoc-glycine ≥98%
N-Fmoc-glycine

Purity: 98% CAS No.: 29022-11-5 Molecular Formula: FmocNHCH2CO2H Formula Weight: 297.32 Storage Temperature: 2°C to 8°C Melting Point: 172-175° Beilstein Reference: 2163967 MDL No.: MFCD00037140

Vzorec: C₁₇H₁₅NO₄
Bod tání: 172…175 °C
MDL Number: MFCD00037140
CAS číslo: 29022-11-5
EINECS: 249-373-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR