Tisk…

3-Aminothiophene-2-carboxamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

632813-1G 632813-250MG
SIAL632813-1GEA 2770 CZK
SIAL632813-1G SIAL632813-250MG
3-Aminothiophene-2-carboxamide, Sigma-Aldrich®
3-Aminothiophene-2-carboxamide
Vzorec: C₅H₆N₂OS
MW: 142,18 g/mol
MDL Number: MFCD00052593
CAS číslo: 147123-47-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR