5-Nitro-2-thiophenecarbonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
5-Nitrothiophene-2-carbonitrile
L11058.03
L11058.06
L11058.03EA
2390
CZK
L11058.03
L11058.06
5-Nitro-2-thiophenecarbonitrile ≥98%
5-Nitro-2-thiophenecarbonitrile
Purity: 98+%
CAS No.: 16689-02-4
Molecular Formula: C5H2N2O2S
Formula Weight: 154.15
Melting Point: 43-45°
Beilstein Reference: 123934
UN No.: 3439
MDL No.: MFCD00052401
Vzorec:
C₅H₂N₂O₂S MW: 154,15 g/mol Bod tání: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052401 CAS číslo: 16689-02-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...