Tisk…

2-Mercaptopyridine-N-oxide sodium salt, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

H3261-100G H3261-10G H3261-1G
SIALH3261-100GEA 46820 CZK
SIALH3261-100G SIALH3261-10G SIALH3261-1G
2-Mercaptopyridine-N-oxide sodium salt, Sigma-Aldrich®
2-Mercaptopyridine-N-oxide sodium salt
Vzorec: C₅H₄NNaOS
MDL Number: MFCD01941547
CAS číslo: 3811-73-2
EINECS: 223-296-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR