Tisk…

2-Mercaptopyridine-N-oxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Mercaptopyridine 1-oxide

188549-25G 188549-5G
SIAL188549-25GEA 7930 CZK
SIAL188549-25G SIAL188549-5G
2-Mercaptopyridine-N-oxide, Sigma-Aldrich®
2-Mercaptopyridine-N-oxide
Vzorec: C₅H₅NOS
MW: 127.17 g/mol
MDL Number: MFCD00006196
CAS číslo: 1121-31-9
EINECS: 214-329-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR