3-Cyanopyridine-N-oxide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-Cyanopyridine 1-oxide, Nicotinonitrile-1-oxide
A19961.88
A19961.09
A19961.88EA
1480
CZK
A19961.88
A19961.09
3-Cyanopyridine-N-oxide ≥97%
3-Cyanopyridine-N-oxide
Purity: 97%
CAS No.: 14906-64-0
Molecular Formula: C6H4N2O
Formula Weight: 120.11
Melting Point: 175-178°
MDL No.: MFCD00129037
Vzorec:
C₆H₄N₂O Bod tání: 175…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00129037 CAS číslo: 14906-64-0 EINECS: 238-975-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...