1-Bromo-4-(dimethylamino)naphthalene 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L17666.03
L17666.06
L17666.03EA
2340
CZK
L17666.03
L17666.06
1-Bromo-4-(dimethylamino)naphthalene 95%
1-Bromo-4-(dimethylamino)naphthalene
Purity: 95%
CAS No.: 59557-93-6
Molecular Formula: C12H12BrN
Formula Weight: 250.14
Refractive Index: 1.6361
Boiling Point: 137-139°/2mm
UN No.: 2810
MDL No.: MFCD02093945
Vzorec:
C₁₂H₁₂BrN MW: 250,14 g/mol Bod varu: 137…139 °C (2 mmHg) Hustota: 1,4305 Storage Temperature: Ambient |
MDL Number:
MFCD02093945 CAS číslo: 59557-93-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...