Tris(trimethylsilyl)borate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tris(trimethylsiloxy)borate
M-Clarity™ quality level = MQ100
348635-100G
348635-25G
SIAL348635-100GEA
11720
CZK
SIAL348635-100G
SIAL348635-25G
Tris(trimethylsilyl)borate, Sigma-Aldrich®
Tris(trimethylsilyl)borate
Vzorec:
C₉H₂₇BO₃Si₃ MW: 278,376 g/mol |
MDL Number:
MFCD00051588 CAS číslo: 4325-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...