Tris(trimethylsilyl)borate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tris(trimethylsiloxy)borate
L06277.06
L06277.14
L06277.06EA
1010
CZK
L06277.06
L06277.14
Tris(trimethylsilyl)borate ≥98%
Tris(trimethylsilyl)borate
Purity: 98%
CAS No.: 4325-85-3
Molecular Formula: [(CH3)3SiO]3B
Formula Weight: 278.38
Flash Point: 42°(107°F)
Refractive Index: 1.3861
Melting Point: -35°
Boiling Point: 183-185°
Beilstein Reference: 1772733
UN No.: 1993
MDL No.: MFCD00051588
Vzorec:
C₉H₂₇BO₃Si₃ MW: 278,376 g/mol Bod varu: 183…185 °C Bod tání: –35 °C Hustota: 0,834 Bod vzplanutí: 42 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051588 CAS číslo: 4325-85-3 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...