Tisk…

Calcium wolframate ≥98%

Dodavatel: Thermo Scientific

Synonyms: Calcium tungstate, Calcium tungsten oxide

13051.14 13051.22 13051.36
13051.14EA 2200 CZK
13051.14 13051.22 13051.36
Calcium wolframate ≥98%
Calcium wolframate

Purity: 98% CAS No.: 7790-75-2 Molecular Formula: CaWO4 Formula Weight: 287.93 Physical Form: -325 Mesh Powder Melting Point: 1620° MDL No.: MFCD00010913

Vzorec: CaO₄W
MW: 287,92 g/mol
Bod tání: 1620 °C
Storage Temperature: Ambient
MDL Number: MFCD00010913
CAS číslo: 7790-75-2
EINECS: 232-219-4
Merck Index: 14,1712

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR