Tisk…

Sodium dodecatungstophosphate hydrate

Dodavatel: Thermo Scientific
Varování

A11575.14 A11575.22 A11575.36
A11575.14EA 2020 CZK
A11575.14 A11575.22 A11575.36
Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate

CAS No.: 51312-42-6 Molecular Formula: Na3PO4.12WO3.xH2O Formula Weight: 2946.12(anhy) Melting Point: 350° MDL No.: MFCD00151658

Vzorec: Na₃O₄₀PW₁₂·H₂O
MW: 2946,12 g/mol (anhy)
Bod tání: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD00151658
CAS číslo: 51312-42-6
EINECS: 257-132-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR