Tisk…

Cyclopropylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

597988-1G 597988-5G
SIAL597988-1GEA 2720 CZK
SIAL597988-1G SIAL597988-5G
Cyclopropylboronic acid, Sigma-Aldrich®
Cyclopropylboronic acid
Vzorec: C₃H₇BO₂
MDL Number: MFCD04038750
CAS číslo: 411235-57-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR