Tisk…

Cyclopropylboronic acid pinacol ester 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H27642.06 H27642.03
H27642.06EA 4770 CZK
H27642.06 H27642.03
Cyclopropylboronic acid pinacol ester 96%
Cyclopropylboronic acid pinacol ester

Purity: 96% CAS No.: 126689-01-8 Molecular Formula: C9H17BO2 Formula Weight: 168.04 Storage Temperature: -30°C to -10°C Flash Point: 40°(104°F) Boiling Point: 146° UN No.: 3272 MDL No.: MFCD05663847

Vzorec: C₉H₁₇BO₂
Bod varu: 146 °C
Hustota: 0,922
Bod vzplanutí: 40 °C (104 °F)
MDL Number: MFCD05663847
CAS číslo: 126689-01-8
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR