5-Bromo-3-phenylsalicylic acid
Dodavatel: Cayman Chemical
13379-1
13379-5
13379-10
13379-25
CAYM13379-1EA
2240
CZK
CAYM13379-1
CAYM13379-5
CAYM13379-10
CAYM13379-25
5-Bromo-3-phenylsalicylic acid
5-Bromo-3-phenylsalicylic acid
Vzorec:
C₁₃H₉BrO₃ |
CAS číslo:
99514-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...