Tisk…

3-Bromo-5-phenylsalicylic acid

Dodavatel: Cayman Chemical

Synonyms: NSC 109116, 5-PBSA, AKR1C1-IN-1

13574-1 13574-5 13574-10 13574-25
CAYM13574-1EA 2670 CZK
CAYM13574-1 CAYM13574-5 CAYM13574-10 CAYM13574-25
3-Bromo-5-phenylsalicylic acid
3-Bromo-5-phenylsalicylic acid
Vzorec: C₁₃H₉BrO₃
MW: 293,113 g/mol
MDL Number: MFCD18378258
CAS číslo: 4906-68-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR