4-Hydroxy-2-(trifluoromethyl)benzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
H32158.MD
H32158.03
H32158.MDEA
2550
CZK
H32158.MD
H32158.03
4-Hydroxy-2-(trifluoromethyl)benzonitrile ≥98%
4-Hydroxy-2-(trifluoromethyl)benzonitrile
Purity: 98%
CAS No.: 320-42-3
Molecular Formula: C8H4F3NO
Formula Weight: 187.12
Melting Point: 120-124°
MDL No.: MFCD06797340
Vzorec:
C₈H₄F₃NO MW: 187,12 g/mol Bod tání: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797340 CAS číslo: 320-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...