Tisk…

Tolbutamide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B21698.09 B21698.18 B21698.30
B21698.09EA 1390 CZK
B21698.09 B21698.18 B21698.30
Tolbutamide ≥98%
Tolbutamide

Purity: 98% CAS No.: 64-77-7 Molecular Formula: C12H18N2O3S Formula Weight: 270.35 Melting Point: 128-130° Beilstein Reference: 1984428 Merck Reference: 14,9507 MDL No.: MFCD00027169

Vzorec: C₁₂H₁₈N₂O₃S
Bod tání: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00027169
CAS číslo: 64-77-7
EINECS: 200-594-3
Merck Index: 14,9507

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR