Tisk…

5-Acetylsalicylic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Acetyl-2-hydroxybenzoic acid, 5-Acetylsalicylic Acid

A11663.06 A11663.14 A11663.22
A11663.06EA 2170 CZK
A11663.06 A11663.14 A11663.22
5-Acetylsalicylic acid ≥98%
5-Acetylsalicylic acid

Purity: 98+% CAS No.: 13110-96-8 Molecular Formula: C9H8O4 Formula Weight: 180.15 Flash Point: 250°(482°F) Melting Point: 214-216° Beilstein Reference: 2096968 MDL No.: MFCD00013978

Vzorec: C₉H₈O₄
MW: 180,15 g/mol
Bod tání: 214…216 °C
Bod vzplanutí: 250 °C (482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013978
CAS číslo: 13110-96-8
EINECS: 236-037-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR