Methyl-5-acetylsalicylate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl 5-acetyl-2-hydroxybenzoate
A10215.14
A10215.22
A10215.14EA
2190
CZK
A10215.14
A10215.22
Methyl-5-acetylsalicylate ≥98%
Methyl-5-acetylsalicylate
Purity: 98%
CAS No.: 16475-90-4
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 58-62°
Beilstein Reference: 2643824
Vzorec:
C₁₀H₁₀O₄ MW: 194,19 g/mol Bod tání: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040924 CAS číslo: 16475-90-4 EINECS: 240-532-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...