4-(Hydroxymethyl)phenylboronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Hydroxymethyl)benzeneboronic acid
L15194.03
L15194.06
L15194.14
L15194.03EA
1750
CZK
L15194.03
L15194.06
L15194.14
4-(Hydroxymethyl)phenylboronic acid ≥98%
4-(Hydroxymethyl)phenylboronic acid
Purity: 98%
CAS No.: 59016-93-2
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: ca 280°
Beilstein Reference: 2831413
Vzorec:
C₇H₉BO₃ MW: 151,96 g/mol Bod tání: ∼ 280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792672 CAS číslo: 59016-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...