3-(Hydroxymethyl)phenylboronic acid pinacol ester
Dodavatel: Thermo Scientific
Synonyms:
2-[3-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hydroxymethylphenylboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 3-(Hydroxymethyl)phenylboronic acid Pinacol ester, 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
H51126.03
H51126.06
H51126.14
H51126.03EA
1530
CZK
H51126.03
H51126.06
H51126.14
3-(Hydroxymethyl)phenylboronic acid pinacol ester
3-(Hydroxymethyl)phenylboronic acid pinacol ester
CAS No.: 443776-76-9
Molecular Formula: C13H19BO3
Formula Weight: 234.10
MDL No.: MFCD09266196
Vzorec:
C₁₃H₁₉BO₃ MW: 234,10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09266196 CAS číslo: 443776-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...