2-Amino-4-(4-cyanophenyl)thiazole 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-(2-Amino-1,3-thiazol-4-yl)benzonitrile
H54861.MD
H54861.03
H54861.06
H54861.MDEA
1310
CZK
H54861.MD
H54861.03
H54861.06
2-Amino-4-(4-cyanophenyl)thiazole 97%
2-Amino-4-(4-cyanophenyl)thiazole
Purity: 97%
CAS No.: 436151-85-8
Molecular Formula: C10H7N3S
Formula Weight: 201.25
Beilstein Reference: 15029305
MDL No.: MFCD03407828
Vzorec:
C₁₀H₇N₃S MW: 201,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03407828 CAS číslo: 436151-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...