Tisk…

Methyl-4-iodsalicylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 2-hydroxy-4-iodobenzoate

M-Clarity™ quality level = MQ100

652636-25G 652636-5G
SIAL652636-25GEA 6410 CZK
SIAL652636-25G SIAL652636-5G
Methyl-4-iodsalicylate, Sigma-Aldrich®
Methyl-4-iodsalicylate
Vzorec: C₈H₇IO₃
MW: 278,05 g/mol
MDL Number: MFCD06797864
CAS číslo: 18179-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR