Tisk…

Methyl thiosalicylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 2-mercaptobenzoate

M-Clarity™ quality level = MQ100

357758-25G 357758-5G
SIAL357758-25GEA 11300 CZK
SIAL357758-25G SIAL357758-5G
Methyl thiosalicylate, Sigma-Aldrich®
Methyl thiosalicylate
Vzorec: C₈H₈O₂S
MW: 168,21 g/mol
MDL Number: MFCD00060678
CAS číslo: 4892-02-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR