Tisk…

Methyl-4-methoxysalicylate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L03855.09 L03855.18
L03855.09EA 1260 CZK
L03855.09 L03855.18
Methyl-4-methoxysalicylate ≥98%
Methyl-4-methoxysalicylate

Purity: 98% CAS No.: 5446-02-6 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 47-51° Boiling Point: 134°/7mm MDL No.: MFCD00008424

Vzorec: C₉H₁₀O₄
Bod varu: 134 °C (7 mmHg)
Bod tání: 47…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008424
CAS číslo: 5446-02-6
EINECS: 226-657-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR