4-(Dimethylamino)benzhydrazide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A13723.09
A13723.18
A13723.09EA
1810
CZK
A13723.09
A13723.18
4-(Dimethylamino)benzhydrazide ≥98%
4-(Dimethylamino)benzhydrazide
Purity: 98+%
CAS No.: 19353-92-5
Molecular Formula: C9H13N3O
Formula Weight: 179.22
Melting Point: 174-175°
Beilstein Reference: 2210856
Vzorec:
C₉H₁₃N₃O MW: 179,22 g/mol Bod tání: 174…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014761 CAS číslo: 19353-92-5 EINECS: 242-985-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...