4-Phenoxybenzonitrile 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B25364.04
B25364.09
B25364.04EA
900
CZK
B25364.04
B25364.09
4-Phenoxybenzonitrile 96%
4-Phenoxybenzonitrile
Purity: 96%
CAS No.: 3096-81-9
Molecular Formula: C13H9NO
Formula Weight: 195.22
Flash Point:
Vzorec:
C₁₃H₉NO MW: 195,22 g/mol Bod tání: 46…48 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017346 CAS číslo: 3096-81-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...