Tisk…

3-Chloro-4-fluorophenylhydrazine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

638374-1G 638374-5G
SIAL638374-1GEA 1500 CZK
SIAL638374-1G SIAL638374-5G
3-Chloro-4-fluorophenylhydrazine, Sigma-Aldrich®
3-Chloro-4-fluorophenylhydrazine
Vzorec: C₆H₆ClFN₂
MDL Number: MFCD00042214
CAS číslo: 84282-78-0
UN: 2811
ADR: 6.1,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR