4-(Diethylamino)salicylaldehyde 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Diethylamino)-2-hydroxybenzaldehyde
L11659.06
L11659.14
L11659.06EA
762
CZK
L11659.06
L11659.14
4-(Diethylamino)salicylaldehyde 99%
4-(Diethylamino)salicylaldehyde
Purity: 99%
CAS No.: 17754-90-4
Molecular Formula: C11H15NO2
Formula Weight: 193.25
Melting Point: 60-64°
Beilstein Reference: 2212234
Vzorec:
C₁₁H₁₅NO₂ MW: 193,25 g/mol Bod tání: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003326 CAS číslo: 17754-90-4 EINECS: 241-745-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...