Tisk…

Methyl-4-formylbenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl terephthalaldehydate, Methyl 4-formylbenzoate

M-Clarity™ quality level = MQ200

244740-25G 244740-5G
SIAL244740-25GEA 10190 CZK
SIAL244740-25G SIAL244740-5G
Methyl-4-formylbenzoate, Sigma-Aldrich®
Methyl-4-formylbenzoate
Vzorec: C₉H₈O₃
MW: 164,16 g/mol
MDL Number: MFCD00006950
CAS číslo: 1571-08-0
EINECS: 216-385-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR