Tisk…

Methyl-3-formylbenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 3-formylbenzoate

47749-1G-F 47749-5G-F
SIAL47749-1G-FEA 2220 CZK
SIAL47749-1G-F SIAL47749-5G-F
Methyl-3-formylbenzoate, Sigma-Aldrich®
Methyl-3-formylbenzoate
Vzorec: C₉H₈O₃
MW: 164,1600 g/mol
MDL Number: MFCD00189379
CAS číslo: 52178-50-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR