5-Bromo-2-iodobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B24880.06
B24880.14
B24880.06EA
3680
CZK
B24880.06
B24880.14
5-Bromo-2-iodobenzonitrile ≥98%
5-Bromo-2-iodobenzonitrile
Purity: 98+%
CAS No.: 121554-10-7
Molecular Formula: C7H3BrIN
Formula Weight: 307.92
Melting Point: 120-122°
Beilstein Reference: 4860282
UN No.: 3276
MDL No.: MFCD00672950
Vzorec:
C₇H₃BrIN MW: 307,92 g/mol Bod tání: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672950 CAS číslo: 121554-10-7 EINECS: 601-788-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...