Tisk…

4-Bromo-2-fluorobenzamide 96%

Dodavatel: Thermo Scientific
Varování

H52336.MD H52336.03
H52336.MDEA 3140 CZK
H52336.MD H52336.03
4-Bromo-2-fluorobenzamide 96%
4-Bromo-2-fluorobenzamide

Purity: 96% CAS No.: 292621-45-5 Molecular Formula: C7H5BrFNO Formula Weight: 218.03 Melting Point: 151-153° MDL No.: MFCD03094432

Vzorec: C₇H₅BrFNO
MW: 218,03 g/mol
Bod tání: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD03094432
CAS číslo: 292621-45-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR