Potassium vinyltrifluoroborate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L17970.03
L17970.06
L17970.03EA
1450
CZK
L17970.03
L17970.06
Potassium vinyltrifluoroborate ≥97%
Potassium vinyltrifluoroborate
Purity: 97%
CAS No.: 13682-77-4
Molecular Formula: C2H3BF3K
Formula Weight: 133.95
Melting Point: 230° dec.
Beilstein Reference: 7777570
Vzorec:
C₂H₃BF₃K Storage Temperature: Ambient |
MDL Number:
MFCD02093335 CAS číslo: 13682-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...