Tisk…

3-Amino-3-(3-chlorophenyl)propionic acid 98%

Dodavatel: Thermo Scientific

Synonyms: 3-Amino-3-(3-chlorophenyl)propanoic acid

L19797.03 L19797.MD
L19797.03EA 2320 CZK
L19797.03 L19797.MD
3-Amino-3-(3-chlorophenyl)propionic acid 98%
3-Amino-3-(3-chlorophenyl)propionic acid

Purity: 98% CAS No.: 68208-21-9 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Storage Temperature: 2°C to 8°C Melting Point: ca 217° dec. Beilstein Reference: 2938807 MDL No.: MFCD00463160

Vzorec: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Bod tání: ∼ 217 °C (rozkl.)
MDL Number: MFCD00463160
CAS číslo: 68208-21-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR