Tisk…

Isopropylboronic acid pinacol ester ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Isopropylboronic acid Pinacol ester, 2-Isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Isopropylboronic acid, pinacol ester

H33861.03 H33861.06 H33861.14
H33861.03EA 803 CZK
H33861.03 H33861.06 H33861.14
Isopropylboronic acid pinacol ester ≥98%
Isopropylboronic acid pinacol ester
Vzorec: C₉H₁₉BO₂
Bod varu: 115 °C (60 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD05663856
CAS číslo: 76347-13-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR