Methanesulphonamide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methylsulphonamide
64275-10G
64275-50G
SIAL64275-10GEA
1880
CZK
SIAL64275-10G
SIAL64275-50G
Methanesulphonamide, Sigma-Aldrich®
Methanesulphonamide
Vzorec:
CH₅NO₂S MW: 95,13 g/mol |
MDL Number:
MFCD00007940 CAS číslo: 3144-09-0 EINECS: 221-553-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...