Methanesulphonamide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methylsulphonamide
A10885.09
A10885.18
A10885.30
A10885.36
A10885.09EA
1070
CZK
A10885.09
A10885.18
A10885.30
A10885.36
Methanesulphonamide ≥98%
Methanesulphonamide
Purity: 98+%
CAS No.: 3144-09-0
Molecular Formula: CH5NO2S
Formula Weight: 95.13
Melting Point: 87-93°
Beilstein Reference: 1740835
Vzorec:
CH₅NO₂S MW: 95,13 g/mol Bod tání: 87…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007940 CAS číslo: 3144-09-0 EINECS: 221-553-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...