Tisk…

Methyl glycolate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hydroxyacetic Acid Methyl Ester, Methyl 2-hydroxyacetate, Glycolic Acid Methyl Ester, Methyl Hydroxyacetate

M-Clarity™ quality level = MQ100

325260-25G 325260-5G
SIAL325260-25GEA 3380 CZK
SIAL325260-25G SIAL325260-5G
Methyl glycolate, Sigma-Aldrich®
Methyl glycolate
Vzorec: C₃H₆O₃
MW: 90,08 g/mol
MDL Number: MFCD00004667
CAS číslo: 96-35-5
EINECS: 202-502-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR